Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTLC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018327.2 | NP_060797.2 | 3 | 725 | ||
XM_011529279.1 | XP_011527581.1 | 3 | 493 | |||
XM_017027922.1 | XP_016883411.1 | 3 | 493 | |||
GenBank mRNA | CF994231.1 | - | 3 | 499 |