siRNA Details
Gene Symbol: TOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080430.3 | NP_001073899.2 | 7 | 2797 | ||
NM_001146188.2 | NP_001139660.1 | 8 | 2799 | |||
XM_005255892.3 | XP_005255949.1 | 7 | 2820 | |||
XM_011523002.2 | XP_011521304.1 | 7 | 3558 | |||
XM_017023142.1 | XP_016878631.1 | 7 | 2873 | |||
GenBank mRNA | AK025084.1 | - | 1 | 1522 | ||
AK095095.1 | - | 2 | 3091 | |||
AK299202.1 | - | 8 | 2799 | |||
U80736.1 | - | 3 | 1898 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
