Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACTIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080543.1 | NP_001074012.1 | 10 | 3557 | ||
NM_021231.1 | NP_067054.1 | 11 | 2625 | |||
GenBank mRNA | AF052087.1 | - | 1 | 1027 | ||
AF155102.1 | - | 3 | 973 | |||
AY917150.1 | - | 12 | 2763 | |||
BC004262.1 | - | 2 | 1779 | |||
BC019848.1 | - | 4 | 1169 |