Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015258.1 | NP_056073.1 | 11 | 1141 | ||
XM_006717018.2 | XP_006717081.1 | 11 | 1058 | |||
XM_006717019.1 | XP_006717082.1 | 10 | 989 | |||
XM_017014552.1 | XP_016870041.1 | 10 | 989 | |||
GenBank mRNA | AB014574.1 | - | 11 | 1068 | ||
AK303074.1 | - | 10 | 989 | |||
AK307498.1 | - | 10 | 970 | |||
AK307507.1 | - | 15 | 1575 | |||
BC009609.1 | - | 11 | 1098 | |||
DA238568.1 | - | 2 | 121 |