Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135995.1 | NP_001129467.1 | 1 | 683 | ||
GenBank mRNA | AB050195.1 | - | 1 | 191 | ||
AK302365.1 | - | 1 | 685 | |||
BC137186.1 | - | 1 | 417 | 9021183 | ||
BC137187.1 | - | 1 | 417 | 9021184 |