Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK074178.1 | - | 10 | 4833 | ||
AK125885.1 | - | 5 | 4166 | |||
AL137619.1 | - | 3 | 2507 |