Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301860.1 | NP_001288789.1 | 20 | 2892 | ||
NR_126051.1 | 20 | 2810 | ||||
XM_011528996.1 | XP_011527298.1 | 20 | 3058 | |||
XM_011528997.2 | XP_011527299.1 | 21 | 3264 | |||
XM_011528998.2 | XP_011527300.1 | 20 | 3081 | |||
XM_011528999.2 | XP_011527301.1 | 21 | 3211 | |||
XM_011529000.1 | XP_011527302.1 | 20 | 3055 | |||
XM_011529001.1 | XP_011527303.1 | 19 | 2737 | |||
XM_011529002.2 | XP_011527304.1 | 19 | 2860 | |||
XM_017028020.1 | XP_016883509.1 | 20 | 2887 | |||
GenBank mRNA | BC140806.1 | - | 20 | 2773 | 9021674 | |
BX647082.1 | - | 17 | 3870 | |||
BX648609.1 | - | 1 | 891 |