Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033602.2 | NP_001028774.2 | 14 | 4492 | ||
NM_015233.5 | NP_056048.1 | 9 | 1544 | |||
XM_011535019.2 | XP_011533321.1 | 17 | 5077 | |||
XM_011535020.2 | XP_011533322.1 | 16 | 5017 | |||
XM_011535021.2 | XP_011533323.1 | 16 | 5014 | |||
XM_011535022.2 | XP_011533324.1 | 14 | 2142 | |||
XM_017020502.1 | XP_016875991.1 | 17 | 5025 | |||
GenBank mRNA | AB018317.2 | - | 14 | 4492 | ||
AK302516.1 | - | 8 | 1235 | |||
BC150245.1 | - | 14 | 4513 | 9007469 |