Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 2 | 278 | ||
NM_001265611.1 | NP_001252540.1 | 2 | 278 | |||
NM_014994.2 | NP_055809.2 | 2 | 278 | |||
NR_049761.1 | 2 | 278 | ||||
NR_049762.1 | 2 | 278 | ||||
XM_006720438.1 | XP_006720501.1 | 2 | 242 | |||
XM_011521383.1 | XP_011519685.1 | 2 | 242 | |||
XM_011521384.2 | XP_011519686.1 | 2 | 242 | |||
XM_017022017.1 | XP_016877506.1 | 1 | 518 | |||
GenBank mRNA | AB011168.2 | - | 2 | 188 | ||
AK091616.1 | - | 2 | 164 | |||
AK292414.1 | - | 2 | 149 | |||
AK302482.1 | - | 2 | 219 | |||
AL833267.1 | - | 2 | 198 | |||
BC036660.1 | - | 2 | 283 | 5266613 | ||
BC113983.1 | - | 2 | 170 | 40083186 | ||
BC114493.1 | - | 2 | 170 | 40083182 | ||
BX648786.1 | - | 2 | 229 |