Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301698.1 | NP_001288627.1 | 6, 7 | 1006 | ||
NM_001301699.1 | NP_001288628.1 | 6, 7 | 638 | |||
NM_001301701.1 | NP_001288630.1 | 6, 7 | 436 | |||
NM_024066.2 | NP_076971.1 | 7, 8 | 1082 | |||
XM_006710891.3 | XP_006710954.1 | 6, 7 | 739 | |||
XM_017002301.1 | XP_016857790.1 | 7, 8 | 1064 | |||
XM_017002303.1 | XP_016857792.1 | 7, 8 | 713 | |||
XM_017002304.1 | XP_016857793.1 | 5, 6 | 698 | |||
XM_017002305.1 | XP_016857794.1 | 5, 6 | 379 | |||
GenBank mRNA | AF007157.1 | - | 7, 8 | 1011 | ||
AK026596.1 | - | 2, 3 | 92 | |||
AK290567.1 | - | 7, 8 | 1070 | |||
AK293833.1 | - | 5, 6 | 426 | |||
AK297708.1 | - | 5, 6 | 628 | |||
BC001072.2 | - | 4, 5 | 296 | |||
BC004456.1 | - | 4, 5 | 296 | |||
BC144384.1 | - | 6, 7 | 980 |