Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015155.2 | NP_055970.1 | 6, 7 | 834 | ||
XM_005252432.3 | XP_005252489.1 | 6, 7 | 843 | |||
XM_005252435.3 | XP_005252492.1 | 6, 7 | 627 | |||
XM_011519435.2 | XP_011517737.1 | 6, 7 | 678 | |||
XM_017015987.1 | XP_016871476.1 | 5, 6 | 2883 | |||
XM_017015988.1 | XP_016871477.1 | 5, 6 | 2881 | |||
XM_017015989.1 | XP_016871478.1 | 5, 6 | 2883 | |||
XM_017015990.1 | XP_016871479.1 | 6, 7 | 2402 | |||
GenBank mRNA | BC131630.1 | - | 5, 6 | 551 | 40118844 | |
BC152443.1 | - | 5, 6 | 558 | 8862568 | ||
D86971.2 | - | 5, 6 | 558 |