Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020802.3 | NP_065853.3 | 6 | 2506 | ||
XM_005271627.4 | XP_005271684.2 | 6 | 2237 | |||
GenBank mRNA | AB037798.1 | - | 3 | 1849 | ||
AK095004.1 | - | 5 | 2136 | |||
AK296350.1 | - | 3 | 2247 |