Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020872.2 | NP_065923.1 | 6 | 760 | ||
XM_005264757.3 | XP_005264814.1 | 7 | 870 | |||
XM_011533768.2 | XP_011532070.1 | 6 | 4392 | |||
XM_017006507.1 | XP_016861996.1 | 7 | 1440 | |||
XM_017006508.1 | XP_016861997.1 | 6 | 665 | |||
GenBank mRNA | AB040929.1 | - | 4 | 409 | ||
BC150608.1 | - | 6 | 760 | 9056978 |