Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC24A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252231.1 | NP_001239160.1 | 8 | 1577 | ||
NM_021982.2 | NP_068817.1 | 8 | 1577 | |||
XM_006714523.2 | XP_006714586.1 | 7 | 1778 | |||
XM_017008961.1 | XP_016864450.1 | 8 | 1222 | |||
XM_017008962.1 | XP_016864451.1 | 8 | 994 | |||
XM_017008963.1 | XP_016864452.1 | 7 | 1778 | |||
GenBank mRNA | AJ131244.1 | - | 8 | 1240 | ||
AK304060.1 | - | 8 | 1566 | |||
BC019341.1 | - | 8 | 1538 | 3355949 |