Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080423.3 | NP_001073892.3 | 18 | 2227 | ||
XM_011534141.2 | XP_011532443.1 | 18 | 2309 | |||
XM_011534142.2 | XP_011532444.1 | 18 | 2221 | |||
XM_011534143.2 | XP_011532445.1 | 17 | 2183 | |||
XM_011534145.2 | XP_011532447.1 | 12 | 1955 | |||
XM_011534146.2 | XP_011532448.1 | 17 | 2788 | |||
GenBank mRNA | AB051506.2 | - | 18 | 2213 | ||
AK024507.1 | - | 5 | 567 |