Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144382.1 | NP_001137854.1 | 2 | 542 | ||
NM_015184.5 | NP_055999.2 | 2 | 336 | |||
XM_006713073.3 | XP_006713136.1 | 2 | 8525 | |||
XM_017006022.1 | XP_016861511.1 | 3 | 767 | |||
XM_017006023.1 | XP_016861512.1 | 3 | 1077 | |||
XM_017006024.1 | XP_016861513.1 | 3 | 696 | |||
XM_017006025.1 | XP_016861514.1 | 3 | 8629 | |||
GenBank mRNA | AB029015.1 | - | 2 | 542 | ||
AK291419.1 | - | 2 | 356 | |||
AK309946.1 | - | 2 | 572 | |||
BC036392.2 | - | 2 | 336 | 5259033 |