Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PM20D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010853.2 | NP_001010853.1 | 7 | 2272 | ||
XM_005248661.2 | XP_005248718.1 | 8 | 2271 | |||
XM_005248662.4 | XP_005248719.1 | 8 | 2022 | |||
XM_011535481.2 | XP_011533783.1 | 7 | 2504 | |||
XM_017010290.1 | XP_016865779.1 | 7 | 2094 | |||
XM_017010291.1 | XP_016865780.1 | 8 | 1914 | |||
GenBank mRNA | AK127529.1 | - | 5 | 1568 | ||
BC035036.1 | - | 7 | 2274 | 5262128 | ||
BC045583.1 | - | 7 | 2022 | |||
BX640865.1 | - | 2 | 1066 |