Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040118.2 | NP_001035207.1 | 7, 8 | 1217 | ||
NM_001135190.1 | NP_001128662.1 | 5, 6 | 1028 | |||
NM_015242.4 | NP_056057.2 | 5, 6 | 1028 | |||
GenBank mRNA | AB018325.2 | - | 5, 6 | 1005 | ||
AF411983.1 | - | 1, 2 | 86 | |||
AJ621557.1 | - | 5, 6 | 1028 | |||
AK160383.1 | - | 4, 5 | 1366 | |||
AY049732.1 | - | 2, 3 | 865 | |||
AY553630.1 | - | 7, 8 | 1232 | |||
BC140792.1 | - | 5, 6 | 698 | 9021660 | ||
BC144562.1 | - | 5, 6 | 698 | 9053090 |