Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BICD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003800.1 | NP_001003800.1 | 1 | 260 | ||
NM_015250.3 | NP_056065.1 | 1 | 260 | |||
XM_017014551.1 | XP_016870040.1 | 1 | 300 | |||
GenBank mRNA | AB014599.1 | - | 1 | 180 | ||
AY052562.1 | - | 1 | 260 | |||
BC073970.1 | - | 1 | 275 | 6169195 |