Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015205.2 | NP_056020.2 | 30 | 4403 | ||
NM_032189.3 | NP_115565.3 | 29 | 4376 | |||
XM_005268299.4 | XP_005268356.1 | 29 | 4674 | |||
XM_005268300.4 | XP_005268357.1 | 29 | 4610 | |||
XM_005268305.4 | XP_005268362.1 | 30 | 4760 | |||
XM_005268306.4 | XP_005268363.1 | 30 | 4757 | |||
GenBank mRNA | AB028944.2 | - | 29 | 4283 | ||
AK024264.1 | - | 3 | 1166 | |||
AK091253.1 | - | 1 | 1805 | |||
AL161996.1 | - | 13 | 2449 | |||
BC035215.1 | - | 1 | 1125 |