Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000697.2 | NP_000688.2 | 1, 2 | 196 | ||
XM_011523780.2 | XP_011522082.1 | 1, 2 | 2470 | |||
GenBank mRNA | BC069557.1 | - | 1, 2 | 174 | 7262463 | |
M35418.1 | - | 1, 2 | 184 | |||
M58704.1 | - | 1, 2 | 176 | |||
M62982.1 | - | 1, 2 | 196 |