Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020884.4 | NP_065935.3 | 22, 23 | 2491 | ||
XM_006723840.3 | XP_006723903.1 | 20, 21 | 2132 | |||
XM_011528941.2 | XP_011527243.1 | 20, 21 | 2156 | |||
XM_011528947.2 | XP_011527249.1 | 14, 15 | 2529 | |||
XM_011528948.2 | XP_011527250.1 | 14, 15 | 1688 | |||
XM_017027986.1 | XP_016883475.1 | 21, 22 | 2192 | |||
XM_017027987.1 | XP_016883476.1 | 16, 17 | 1707 | |||
GenBank mRNA | AB040945.2 | - | 20, 21 | 2200 | ||
BC151242.1 | - | 20, 21 | 2212 | 8860515 |