Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PHACTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100164.1 | NP_001093634.1 | 8 | 1671 | ||
NM_001100165.1 | NP_001093635.1 | 7 | 1431 | |||
NM_001100166.1 | NP_001093636.1 | 7 | 1329 | |||
NM_014721.2 | NP_055536.2 | 8 | 1569 | |||
GenBank mRNA | AB014580.1 | - | 8 | 1486 | ||
AK297799.1 | - | 8 | 1476 | |||
AK298263.1 | - | 8 | 1671 | |||
AK300221.1 | - | 7 | 1324 | |||
BC150630.1 | - | 8 | 1476 | 9057000 | ||
BC152416.1 | - | 8 | 1486 | 8862521 | ||
CR749345.1 | - | 7 | 1369 |