Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080428.2 | NP_001073897.2 | 19 | 4182 | ||
NM_001122679.1 | NP_001116151.1 | 23 | 4754 | |||
XM_005265950.2 | XP_005266007.1 | 25 | 5401 | |||
XM_005265952.2 | XP_005266009.1 | 21 | 4128 | |||
XM_006714897.2 | XP_006714960.1 | 24 | 5380 | |||
XM_011534604.2 | XP_011532906.1 | 24 | 5215 | |||
XM_017009660.1 | XP_016865149.1 | 25 | 5428 | |||
XM_017009661.1 | XP_016865150.1 | 24 | 5407 | |||
XM_017009662.1 | XP_016865151.1 | 24 | 5218 | |||
XM_017009663.1 | XP_016865152.1 | 22 | 4623 | |||
XM_017009664.1 | XP_016865153.1 | 22 | 4536 | |||
XM_017009665.1 | XP_016865154.1 | 22 | 4332 | |||
XM_017009666.1 | XP_016865155.1 | 21 | 4311 | |||
XM_017009667.1 | XP_016865156.1 | 22 | 4305 | |||
XM_017009668.1 | XP_016865157.1 | 22 | 5842 | |||
XM_017009669.1 | XP_016865158.1 | 22 | 5674 | |||
GenBank mRNA | AB032953.2 | - | 14 | 2891 | ||
BX648178.1 | - | 18 | 3776 | |||
CR936703.1 | - | 19 | 4115 |