Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014931.3 | NP_055746.3 | 4, 5 | 801 | ||
GenBank mRNA | AB029038.2 | - | 4, 5 | 1126 | ||
AK297152.1 | - | 4, 5 | 660 | |||
AL544921.3 | - | 4, 5 | 808 | |||
BC002799.2 | - | 3, 4 | 403 | |||
BC068014.1 | - | 3, 4 | 465 | |||
BC094753.1 | - | 4, 5 | 663 | 4838365 | ||
BX362839.2 | - | 3, 4 | 501 |