Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPGAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320808.1 | NP_001307737.1 | 8 | 2967 | ||
NM_014873.2 | NP_055688.1 | 8 | 2378 | |||
XM_005273362.3 | XP_005273419.1 | 9 | 2526 | |||
XM_011510228.2 | XP_011508530.1 | 9 | 2415 | |||
XM_011510229.2 | XP_011508531.1 | 8 | 2298 | |||
XM_017003004.1 | XP_016858493.1 | 7 | 3530 | |||
GenBank mRNA | D86960.1 | - | 8 | 2378 |