Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130960.1 | NP_001124432.1 | 22 | 3129 | ||
NM_001130961.1 | NP_001124433.1 | 22 | 3129 | |||
NM_014996.2 | NP_055811.1 | 21 | 2812 | |||
XM_005247238.1 | XP_005247295.1 | 23 | 3178 | |||
XM_005247239.1 | XP_005247296.1 | 21 | 3169 | |||
XM_011512560.2 | XP_011510862.1 | 23 | 3187 | |||
XM_011512561.2 | XP_011510863.1 | 23 | 3319 | |||
XM_011512562.2 | XP_011510864.1 | 24 | 3936 | |||
XM_011512564.2 | XP_011510866.1 | 22 | 3118 | |||
XM_011512565.2 | XP_011510867.1 | 22 | 3650 | |||
XM_011512566.2 | XP_011510868.1 | 22 | 3291 | |||
XM_011512567.1 | XP_011510869.1 | 23 | 3178 | |||
XM_017005923.1 | XP_016861412.1 | 23 | 3178 | |||
XM_017005924.1 | XP_016861413.1 | 22 | 3118 | |||
XM_017005925.1 | XP_016861414.1 | 22 | 3106 | |||
XM_017005926.1 | XP_016861415.1 | 22 | 3077 | |||
XM_017005927.1 | XP_016861416.1 | 23 | 3178 | |||
XM_017005928.1 | XP_016861417.1 | 23 | 3178 | |||
XM_017005929.1 | XP_016861418.1 | 22 | 3229 | |||
GenBank mRNA | AB028992.1 | - | 18 | 2207 | ||
AK022610.1 | - | 13 | 1621 | |||
AK096620.1 | - | 21 | 3169 | |||
AY691170.1 | - | 22 | 2852 | |||
AY691171.1 | - | 22 | 2852 | |||
BC043248.2 | - | 1 | 78 | |||
BC113950.1 | - | 21 | 2812 | 5501480 |