Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039613.2 | NP_001034702.1 | 6 | 838 | ||
NM_001320858.1 | NP_001307787.1 | 6 | 938 | |||
NM_001320859.1 | NP_001307788.1 | 5 | 785 | |||
NM_001320860.1 | NP_001307789.1 | 5 | 753 | |||
NM_001320863.1 | NP_001307792.1 | 6 | 806 | |||
XM_017003878.1 | XP_016859367.1 | 5 | 928 | |||
GenBank mRNA | AK291984.1 | - | 6 | 805 | ||
AK297648.1 | - | 5 | 739 | |||
BC019061.1 | - | 5 | 777 | |||
BC039709.1 | - | 6 | 805 | |||
BC059410.1 | - | 5 | 760 | |||
BC067865.1 | - | 5 | 749 | |||
BC111025.1 | - | 6 | 826 | 5200514 |