Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025153.2 | NP_079429.2 | 8 | 1537 | ||
XM_006714833.2 | XP_006714896.1 | 7 | 1118 | |||
XM_011534468.2 | XP_011532770.1 | 7 | 1094 | |||
XM_011534469.1 | XP_011532771.1 | 8 | 1358 | |||
XM_011534472.2 | XP_011532774.1 | 3 | 834 | |||
XM_017009252.1 | XP_016864741.1 | 7 | 1094 | |||
XM_017009253.1 | XP_016864742.1 | 7 | 1118 | |||
GenBank mRNA | AB018258.2 | - | 8 | 1364 | ||
AK025130.1 | - | 3 | 646 | |||
AK090832.1 | - | 7 | 1353 | |||
AK295081.1 | - | 6 | 1203 |