Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277285.1 | NP_001264214.1 | 13 | 1793 | ||
XM_006718795.3 | XP_006718858.1 | 2 | 307 | |||
XM_011542691.2 | XP_011540993.1 | 14 | 2480 | |||
XM_011542692.2 | XP_011540994.1 | 14 | 2468 | |||
XM_011542694.2 | XP_011540996.1 | 14 | 2483 | |||
XM_011542695.2 | XP_011540997.1 | 13 | 2011 | |||
XM_017017393.1 | XP_016872882.1 | 13 | 2498 | |||
XM_017017394.1 | XP_016872883.1 | 14 | 2483 | |||
XM_017017395.1 | XP_016872884.1 | 14 | 2482 | |||
XM_017017396.1 | XP_016872885.1 | 7 | 1822 | |||
GenBank mRNA | AB028953.1 | - | 1 | 35 | ||
AK097578.1 | - | 10 | 1523 |
Set Membership: |
G-protein coupled receptor |