Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277285.1 | NP_001264214.1 | 16 | 2187 | ||
XM_006718795.3 | XP_006718858.1 | 5 | 701 | |||
XM_011542691.2 | XP_011540993.1 | 17 | 2874 | |||
XM_011542692.2 | XP_011540994.1 | 17 | 2862 | |||
XM_011542694.2 | XP_011540996.1 | 17 | 2877 | |||
XM_011542695.2 | XP_011540997.1 | 16 | 2405 | |||
XM_017017393.1 | XP_016872882.1 | 16 | 2892 | |||
XM_017017394.1 | XP_016872883.1 | 17 | 2877 | |||
XM_017017395.1 | XP_016872884.1 | 17 | 2876 | |||
XM_017017396.1 | XP_016872885.1 | 10 | 2216 | |||
GenBank mRNA | AB028953.1 | - | 4 | 429 | ||
AK097578.1 | - | 13 | 1917 |
Set Membership: |
G-protein coupled receptor |