Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020410.2 | NP_065143.2 | 24 | 3291 | ||
GenBank mRNA | AB058728.1 | - | 24 | 3261 | ||
AF288687.1 | - | 24 | 2909 | |||
AK026044.1 | - | 8 | 1441 | |||
AK056420.1 | - | 24 | 3267 | |||
AK074159.1 | - | 15 | 4076 | |||
AK074798.1 | - | 12 | 1796 | |||
AK074920.1 | - | 13 | 1871 | |||
AK095287.1 | - | 7 | 2464 | |||
AK123780.1 | - | 7 | 1635 | |||
AK125861.1 | - | 8 | 3588 | |||
AK130125.1 | - | 15 | 1931 | |||
AK172778.1 | - | 23 | 2722 | |||
AL834180.1 | - | 1 | 1740 | |||
BC009302.2 | - | 6 | 735 | |||
BC069211.1 | - | 5 | 625 |