Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022153.1 | NP_071436.1 | 7 | 1435 | ||
GenBank mRNA | AF193048.1 | - | 2 | 574 | ||
AF289594.1 | - | 1871 | ||||
AK024449.1 | - | 7 | 1353 | |||
AY265805.1 | - | 7 | 1350 | |||
AY358379.1 | - | 7 | 1435 | |||
BC020568.1 | - | 7 | 1307 | 4158410 | ||
BC111048.1 | - | 7 | 1283 | |||
EF506490.1 | - | 7 | 1347 |