Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015082.1 | NP_055897.1 | 13, 14 | 1832 | ||
XM_011543283.1 | XP_011541585.1 | 13, 14 | 1739 | |||
XM_011543284.1 | XP_011541586.1 | 12, 13 | 1783 | |||
XM_011543286.2 | XP_011541588.1 | 10, 11 | 1293 | |||
XM_011543287.2 | XP_011541589.1 | 7, 8 | 1521 | |||
XM_017009251.1 | XP_016864740.1 | 8, 9 | 1548 | |||
GenBank mRNA | AB028984.1 | - | 9, 10 | 1169 | ||
AK056284.1 | - | 5, 6 | 832 | |||
BC024300.1 | - | 13, 14 | 1864 | 4662520 | ||
BX640845.1 | - | 11, 12 | 1247 |