Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBSCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU180714.1 | - | 196 |
Gene Symbol: C1orf145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073154.1 | 1 | 206 | |||
NR_073155.1 | 1 | 197 | ||||
GenBank mRNA | AK056556.1 | - | 1 | 206 | ||
BC027909.1 | - | 1 | 197 |