Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152432.2 | NP_689645.2 | 17, 18 | 1550 | ||
XM_011542615.2 | XP_011540917.1 | 17, 18 | 4038 | |||
XM_011542616.2 | XP_011540918.1 | 17, 18 | 1807 | |||
XM_011542617.2 | XP_011540919.1 | 18, 19 | 1693 | |||
XM_017017237.1 | XP_016872726.1 | 17, 18 | 1996 | |||
XM_017017238.1 | XP_016872727.1 | 18, 19 | 4179 | |||
GenBank mRNA | AK057372.1 | - | 5, 6 | 420 | ||
BU146672.1 | - | 1, 2 | 56 |