Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152432.2 | NP_689645.2 | 1, 2 | 166 | ||
GenBank mRNA | AI672473.1 | - | 1, 2 | 167 |