Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRTG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173814.5 | NP_776175.2 | 18, 19 | 3289 | ||
XM_017022081.1 | XP_016877570.1 | 18, 19 | 3376 | |||
GenBank mRNA | AK098622.1 | - | 9, 10 | 1297 | ||
AY630258.1 | - | 18, 19 | 3149 |