Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080463.1 | NP_001073932.1 | 34 | 5308 | ||
NM_001377.2 | NP_001368.2 | 34 | 5308 | |||
XM_006718903.2 | XP_006718966.1 | 34 | 5308 | |||
XM_017018291.1 | XP_016873780.1 | 34 | 5308 | |||
XM_017018292.1 | XP_016873781.1 | 31 | 4562 | |||
XM_017018293.1 | XP_016873782.1 | 34 | 5308 | |||
GenBank mRNA | AB290167.1 | - | 34 | 5362 | ||
U20552.1 | - | 2 | 154 | |||
U53531.1 | - | 13 | 1896 | |||
Z83800.1 | - | 2 | 100 |