Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002880.1 | NP_001002880.2 | 5 | 539 | ||
NM_015373.3 | NP_056188.1 | 4 | 399 | |||
GenBank mRNA | AB015347.1 | - | 1 | 86 | ||
AB111855.1 | - | 3 | 277 | |||
AF331041.1 | - | 4 | 335 | |||
AK307265.1 | - | 3 | 494 | |||
AK311928.1 | - | 4 | 412 | |||
AL050345.1 | - | 4 | 378 | |||
AL136686.1 | - | 4 | 382 | |||
BC016139.1 | - | 4 | 377 | 3922707 | ||
CR456410.1 | - | 3 | 291 |