Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 30 | 4735 | ||
XM_006711956.2 | XP_006712019.1 | 30 | 4840 | |||
XM_011532649.2 | XP_011530951.1 | 30 | 4689 | |||
XM_011532650.2 | XP_011530952.1 | 30 | 4690 | |||
XM_011532652.1 | XP_011530954.1 | 29 | 4609 | |||
XM_011532653.1 | XP_011530955.1 | 30 | 4735 | |||
XM_011532654.1 | XP_011530956.1 | 29 | 4468 | |||
XM_011532655.2 | XP_011530957.1 | 26 | 3757 | |||
XM_011532657.1 | XP_011530959.1 | 24 | 3496 | |||
XM_011532660.1 | XP_011530962.1 | 30 | 4735 | |||
XM_017003521.1 | XP_016859010.1 | 29 | 4609 | |||
XM_017003522.1 | XP_016859011.1 | 25 | 4561 | |||
XM_017003523.1 | XP_016859012.1 | 25 | 3728 | |||
XM_017003524.1 | XP_016859013.1 | 30 | 4735 | |||
GenBank mRNA | AJ132086.1 | - | 2 | 173 |