Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034845.2 | NP_001030017.2 | 7 | 1544 | ||
XM_011531993.2 | XP_011530295.1 | 6 | 882 | |||
XM_011531994.1 | XP_011530296.1 | 6 | 716 | |||
XM_011531995.1 | XP_011530297.1 | 5 | 627 | |||
XM_011531996.1 | XP_011530298.1 | 4 | 527 | |||
XM_011531997.1 | XP_011530299.1 | 4 | 449 | |||
XM_017008243.1 | XP_016863732.1 | 7 | 2278 | |||
XM_017008244.1 | XP_016863733.1 | 6 | 1650 | |||
GenBank mRNA | AJ626725.1 | - | 6 | 757 | ||
AK295464.1 | - | 6 | 960 | |||
BC047551.1 | - | 7 | 1465 | 5301536 | ||
GU220060.1 | - | 6 | 808 |