Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145418.1 | NP_001138890.1 | 23 | 6322 | ||
XM_005261405.2 | XP_005261462.1 | 22 | 6298 | |||
XM_006724171.3 | XP_006724234.1 | 22 | 15100 | |||
XM_011530018.2 | XP_011528320.1 | 23 | 6501 | |||
XM_017028673.1 | XP_016884162.1 | 23 | 6295 | |||
GenBank mRNA | AB028966.3 | - | 18 | 5115 | ||
AB665284.1 | - | 23 | 6270 | |||
AL080174.1 | - | 2 | 453 | |||
BC016465.1 | - | 15 | 2866 | 4154050 |