Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 46 | 7388 | ||
XM_006721206.2 | XP_006721269.1 | 46 | 7282 | |||
XM_011523146.2 | XP_011521448.1 | 47 | 7417 | |||
XM_011523147.1 | XP_011521449.1 | 47 | 7384 | |||
XM_011523148.1 | XP_011521450.1 | 47 | 7457 | |||
XM_011523151.1 | XP_011521453.1 | 46 | 7314 | |||
XM_011523152.1 | XP_011521454.1 | 9 | 1141 | |||
XM_011523155.2 | XP_011521457.1 | 2 | 142 | |||
XM_017023346.1 | XP_016878835.1 | 46 | 7560 | |||
XM_017023347.1 | XP_016878836.1 | 34 | 5799 | |||
XM_017023348.1 | XP_016878837.1 | 34 | 5539 |
Gene Symbol: HYDIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103556.1 | 38 | 6144 | |||
GenBank mRNA | AB058767.1 | - | 12 | 1876 |