Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099433.1 | NP_001092903.1 | 15 | 2431 | ||
XM_017007788.1 | XP_016863277.1 | 15 | 2395 | |||
XM_017007789.1 | XP_016863278.1 | 15 | 2219 | |||
XM_017007790.1 | XP_016863279.1 | 15 | 2371 | |||
XM_017007791.1 | XP_016863280.1 | 15 | 2395 | |||
XM_017007792.1 | XP_016863281.1 | 15 | 2395 | |||
XM_017007793.1 | XP_016863282.1 | 15 | 2395 | |||
XM_017007794.1 | XP_016863283.1 | 15 | 2395 | |||
XM_017007795.1 | XP_016863284.1 | 15 | 2395 | |||
XM_017007796.1 | XP_016863285.1 | 15 | 2537 | |||
XM_017007797.1 | XP_016863286.1 | 14 | 1900 | |||
XM_017007798.1 | XP_016863287.1 | 14 | 2538 | |||
XM_017007799.1 | XP_016863288.1 | 14 | 1900 | |||
GenBank mRNA | AM412307.1 | - | 15 | 2395 | ||
AM412310.1 | - | 13 | 1840 |