Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015356.4 | NP_056171.3 | 36 | 5122 | ||
NM_182706.4 | NP_874365.3 | 37 | 5197 | |||
GenBank mRNA | AF240677.1 | - | 36 | 5115 | ||
AF271734.2 | - | 36 | 5093 | |||
AF318352.1 | - | 7 | 1669 | |||
AY062238.1 | - | 36 | 5124 | |||
BC009490.2 | - | 13 | 1577 | |||
BC014632.2 | - | 18 | 2721 | |||
BC044627.1 | - | 16 | 2272 | |||
D63481.3 | - | 36 | 5117 |