Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100128288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024447.1 | 1 | 1003 | |||
GenBank mRNA | AK094860.1 | - | 1 | 682 |