Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015379.4 | NP_056194.1 | 3 | 697 | ||
GenBank mRNA | AB055977.1 | - | 3 | 689 | ||
AF041430.2 | - | 3 | 667 | |||
AF106966.1 | - | 3 | 648 | |||
BC018737.1 | - | 3 | 689 | 4875257 | ||
BC062370.1 | - | 3 | 661 | 6598528 |