Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015380.4 | NP_056195.3 | 9 | 1031 | ||
GenBank mRNA | AF151809.1 | - | 9 | 943 | ||
AK001087.1 | - | 9 | 970 | |||
AK097015.1 | - | 2 | 1646 | |||
AK222657.1 | - | 9 | 905 | |||
AK297770.1 | - | 8 | 714 | |||
AK312773.1 | - | 9 | 866 | |||
AY189688.1 | - | 9 | 946 | |||
BC007830.1 | - | 9 | 938 | 3635175 | ||
BC011681.2 | - | 9 | 991 | 3010765 | ||
BC015200.1 | - | 9 | 881 | 4333051 | ||
CR456483.1 | - | 9 | 843 |