Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015155.2 | NP_055970.1 | 18 | 5595 | ||
XM_017015987.1 | XP_016871476.1 | 17 | 7749 | |||
XM_017015988.1 | XP_016871477.1 | 17 | 7642 | |||
XM_017015989.1 | XP_016871478.1 | 17 | 7687 | |||
XM_017015992.1 | XP_016871481.1 | 13 | 4963 | |||
XM_017015993.1 | XP_016871482.1 | 16 | 5349 | |||
XM_017015994.1 | XP_016871483.1 | 17 | 5281 | |||
GenBank mRNA | BC003381.1 | - | 1 | 252 | ||
BC152443.1 | - | 17 | 5319 | 8862568 | ||
D86971.2 | - | 17 | 5319 |